Supelco® Organic Reference Standard, ᴅ-(+)-Raffinose pentahydrate

Toimittaja: Merck

95068-100MG
95068-100MGEA 66.4 EUR
95068-100MG
Supelco® Organic Reference Standard, ᴅ-(+)-Raffinose pentahydrate
Standardiaineet ja vertailumateriaalit Organic Standards

Order Now

TEKNISET TIEDOT

Synonym(s) O-α-ᴅ-Galactopyranosyl-(1→6)-α-ᴅ-glucopyranosyl β-ᴅ-fructofuranoside; Melitose; Melitriose
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nro 17629-30-0
MDL No. MFCD00071590
Molekyylipaino 594,51
Shipping conditions STD
Säilytysolosuhteet 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR