Reactive dyes and protein labeling kits, CF®
Toimittaja: Biotium
92130 92135 92110 92111 92120 92131 92132 92134 92137 92139 92142 92150 92155 92020 92021 92022 92023 92024 92025 92026 92027 92028 92041 92040 92042 92050 92051 92052 92053 92109 92133 92210 92211 92212 92213 92214 92215 92216 92217 92029 92030 92031 92035 92036 92037 92038 92039 92218 92219 92220 92221 92222 92223 92106 92108 92043 92058 92225 92032 92033 92034 92054 92055 92056 92057 92059 92107 92226 92209 92044 92105 92045 92104 92208 92046 92228 92065 92102 92127 92128 96011 96012 96010 96108 96107
92130.EA 310 EUR
92130. 92135. 92110. 92111. 92120. 92131. 92132. 92134. 92137. 92139. 92142. 92150. 92155. 92020. 92021. 92022. 92023. 92024. 92025. 92026. 92027. 92028. 92041. 92040. 92042. 92050. 92051. 92052. 92053. 92109. 92133. 92210. 92211. 92212. 92213. 92214. 92215. 92216. 92217. 92029. 92030. 92031. 92035. 92036. 92037. 92038. 92039. 92218. 92219. 92220. 92221. 92222. 92223. 92106. 92108. 92043. 92058. 92225. 92032. 92033. 92034. 92054. 92055. 92056. 92057. 92059. 92107. 92226. 92209. 92044. 92105. 92045. 92104. 92208. 92046. 92228. BTIU92065 92102. 92127. 92128. 96011. 96012. 96010. 96108. 96107.
Reactive dyes and protein labeling kits, CF®
Proteiinien yhdistämis- ja leimausjärjestelmät
Biotium offers a number of reactive dye formats for labelling proteins, nucleic acids, and other biomolecules with fluorescent dyes. CF® dyes offer advantages in brightness and photostability compared other similar commercial dyes. CF® dyes are available in a number of reactive dye formats for CF® dye colours span the visible and near-infrared spectra.
Amine groups react with activated carboxylic acids. Aminooxy (also known as hydroxylamine) groups readily react with aldehydes or ketones without the need for a reducing agent. Hydrazides can be microinjected into cells as fixable polar tracers for visualisation of neuronal morphology. Hydrazides also can be used for labelling aldehyde or ketone groups, such as carbohydrate molecules after peroxidation with periodate. However, for this application it is recommended that CF® dye aminooxy forms are used, which are more reactive toward these groups than hydrazide forms. Maleimide reactive dyes label thiols, such as cysteine residues in proteins, and can be used as an alternative to SE labelling in cases where lysine modification alters the biological activity of the labelled protein. Succinimidyl ester (SE) groups label free amines, such as lysine residues in proteins, and are commonly used to label antibodies. CF® dye SE groups are more resistant to hydrolysis compared to most commercially available SE dyes, resulting in improved stability and labelling efficiency.
CF® dye technologies are covered by pending US and international patents. CF® is a registred of Biotium, Inc.
Toimitustiedot: Succinimidyl ester, maleimide and SE protein labelling kits include a detailed protocol for labelling IgG or other proteins. SE Protein Labeling Kits include CF® dye SE and other components required to label and purify proteins. The kits are sufficient for three labelling reactions of 1 mg protein each.
Order Now
TEKNISET TIEDOT
Environmentally Preferable
Isäntä