Kloroformi ≥99.8% (by GC, corrected for water content), puhdistettu, J.T.Baker®

Toimittaja: Avantor
Vaara

9182-01EA 76.86 EUR
9182-01 9182-03
Kloroformi ≥99.8% (by GC, corrected for water content), puhdistettu, J.T.Baker®
Kloroformi

Purified - Chemicals of good quality where there are nofficial standards or compendia, or in some cases, that meet the requirements of former compendia articles.

Kaava: CHCl₃
Kiehumispiste: 61,7 °C (1013 hPa)
Sulamispiste: –63 °C
Tiheys: 1,4832 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000826
CAS-numero: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

For Laboratory,Research,or Manufacturing Use = 99.8 %
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Specific Gravity at 25°/25°C 1.476 - 1.480
Nonvolatile Residue (w/v) ≤0.002 %
Free Chlorine Passes Test
Readily Carbonizable Substances Passes Test
Chlorinated Decomposition Products and Chloride Passes Test
Acid and Phosgene Passes Test
Aldehydes and Ketones (as HCHO) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR