Rikkihiili ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor
Vaara

9172-22EA 234 EUR
9172-22 9172-05
Rikkihiili ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Rikkihiili

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Kaava: CS₂
Kiehumispiste: 46,2 °C (1013 hPa)
Sulamispiste: –111,6 °C
Tiheys: 1,2632 g/cm³ (20 °C)
Leimahduspiste: –30 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011321
CAS-numero: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets ACS Reagent Chemical Requirements = 99.9 %
For Laboratory,Research,or Manufacturing Use = 10
Assay (CS₂) (by GC, corrected for water) ≥99.9 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Hydrogen Sulfide (H₂S) Passes Test
Sulfur Dioxide (SO₂) Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR