VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Toimittaja: VWR Chemicals
91027.050EA 80.4 EUR
91027.050 91028.050 91031.050 91031.010 91027.010 91072.050 91073.050 91074.010 91074.050 91087.002 91087.010 91087.050 91088.001 91088.010 91088.050 91090.001 91090.010 91090.050 91091.001 91091.002 91091.010 91072.010 91250.001
VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Standardiaineet ja vertailumateriaalit Kaasukromatografiastandardit
Avantor offers an extensive range of FAME standards as Certified Reference Materials under the well-known PESTINORM® brand.
  • All products are CRM acc ISO/IEC 17025 and ISO 17034
  • All products are NEAT
  • 24 month shelf life
Esterification of fatty acids is an important tool to determine content and characterise fats and oils in food, especially for measurement by GC methods. The simple derivatisation to create the methyl ester analogue is also used widely as a preliminary technique for assessing the quality and purity of biofuels. VWR® offers a comprehensive range of fatty acid methyl esters (FAME) Certified Reference Materials for calibration of GC instruments, for validation of analytical methods, preparation of 'working reference samples' or for detection limit and linearity studies.
Sertifikaatit: ISO 17025 / 17034