Biological indicators, ampoules for steam sterilisation, STERIL CONTROL
Toimittaja: LIOFILCHEM
91050 91040 91052 91054
91050-VWREA 217 EUR
91050-VWR 91040. 91052-VWR 91054-VWR
Biological indicators, ampoules for steam sterilisation, STERIL CONTROL
Sterilointi-indikaattori
Biological indicators for validating sterilisation using steam sterilisation (autoclaves).
Ampoules with nutrient medium, pH indicator and spores of Geobacillus stearothermophilus (GST).