Supelco® TraceCERT® Organic Reference Standard, ʟ-Methionine sulfoximine
Toimittaja: Merck
91016-100MG
91016-100MGEA 280 EUR
91016-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Methionine sulfoximine
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7c4488f8baf642e0a46a003db9c7b6e41882076&vid=c7c4488f8baf642e0a46a003db9c7b6e41882076&prid=web&clid=SAPDAM