2-Propanoli ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®

Toimittaja: Avantor

Synonyms: Isopropyylialkoholi

Management of Change (MOC) category = N

9084-03EA 305.49 EUR
9084-03 9084-01 9084-R 9084-33 9084-07
2-Propanoli ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
2-Propanoli
Kaava: (CH₃)₂CHOH
MW: 60.1 g/mol
Kiehumispiste: 82 °C (1013 hPa)
Sulamispiste: –88.5 °C
Tiheys: 0.786 g/cm³ (20 °C)
Leimahduspiste: 12 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011674
CAS-numero: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CHOHCH₃) ≥ 99.5 %
Carbonyl Compounds - Acetone ≤ 0.002 %
Carbonyl Compounds - Propionaldehyde ≤ 0.002 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.001 %
Solubility in H₂O Passes Test
Titrable Acid or Base (meq/g) ≤ 0.0001
Water (by KF, coulometric) ≤ 0.2 %
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR