Supelco® TraceCERT® Organic Reference Standard, Tolylfluanid
Toimittaja: Merck
90619-50MG
90619-50MGEA 207 EUR
90619-50MG
Supelco® TraceCERT® Organic Reference Standard, Tolylfluanid
Standardiaineet ja vertailumateriaalit Organic Standards
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Varoitus: P273 - P280 - P302 + P352 - P304 + P340 + P310 - P305 + P351 + P338 - P314