Supelco® Organic Reference Standard, Bisdemethoxycurcumin

Toimittaja: Merck

90594-10MG
90594-10MGEA 202 EUR
90594-10MG
Supelco® Organic Reference Standard, Bisdemethoxycurcumin
Standardiaineet ja vertailumateriaalit Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dfc4be84898cd71a2da26492db50c21337029a1&vid=1dfc4be84898cd71a2da26492db50c21337029a1&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Synonym(s) (E,E)-1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione; Bisdesmethoxycurcumin; Curcumin III; Didemethoxycurcumin
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nro 33171-05-0
MDL No. MFCD03419284
Molekyylipaino 308,33
Shipping conditions STD
Säilytysolosuhteet 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR