Organic reference standard, 4-Deoxy-D-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Toimittaja: Merck
90061-50MG
90061-50MGEA 0 EUR
90061-50MG
Organic reference standard, 4-Deoxy-D-erythronic acid sodium salt hydrate, ≥97,0% (GC), Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
4-Deoxyerythronic acid has been associated with uremia and diabetes mellitus type 1, but is also a normal organic acid present in human biofluids, derived presumably from L-threonine.