Multi-element ion chromatography anion standard solution, CRM, Supelco®
Toimittaja: Merck
89886-50ML-F
89886-50ML-FEA 300 EUR
89886-50ML-F
Multi-element ion chromatography anion standard solution, CRM, Supelco®
Standardiaineet ja vertailumateriaalit Anionic Standards Multi-Element Anioinic Standards
This standard solution is a certified primary reference material for ion chromatography.
Produced with certified high purity materials (NaF, NaCl, NaBr, NaNO₃, Na₂HPO₄, Na₂SO₄) and ultrapure water (18 MOhm, 0,22 μm filtered).
Sertifikaatit: Traceable to SI.