Tolueeni ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

89681-10L 89681-4X25ML
89681-10LEA 436 EUR
89681-10L 89681-4X25ML
Tolueeni ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Tolueeni

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 635760

Kaava: C₆H₅CH₃
Kiehumispiste: 7,1 %(V)
Sulamispiste: -95 °C
Tiheys: 0,867 g/cm³ (20 °C)
Leimahduspiste: 4 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008512
CAS-numero: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Vapor Pressure 26mmHg(25°C)
Assay (GC) min. 99.7%
Density (D20/20) 0.865-0.869
Refractive index (n20/D) 1.496-1.498
Non-volatilematter max.0.001%
Water (KarlFischer) max.0.02%
Freeacid (asHCl) max.0.001%
Freealkali (asNaOH) max.0.001%
Aluminium (Al) max.0.00005%
Barium (Ba) max.0.00001%
Bismuth (Bi) max.0.00001%
Calcium (Ca) max.0.00005%
Cadmium (Cd) max.0.000005%
Cobalt (Co) max.0.000002%
Chromium (Cr) max.0.000002%
Copper (Cu) max.0.000002%
Iron (Fe) max.0.00001%
Potassium (K) max.0.00005%
Lithium (Li) max.0.00001%
Magnesium (Mg) max.0.00001%
Manganese (Mn) max.0.000002%
Molybdenum (Mo) max.0.00001%
Sodium (Na) max.0.00005%
Nickel (Ni) max.0.000002%
Lead (Pb) max.0.00001%
Strontium (Sr) max.0.00001%
Tin (Sn) max.0.00001%
Zinc (Zn) max.0.00001%
S-compounds (asS) max.0.003%
Thiophene max.0.0001%
Reaction against H₂SO₄ complying
APHA max.10
VaporDensity 3.2(vsair)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR