J.T.Baker® HYDRA-POINT™, Volumetric Reagents for Water Determination by Karl Fischer titration
Toimittaja: Avantor
8855.1000EA 91.12 EUR
8855.1000 8792.1000 8792.2500 8844.1000 8844.2500 8855.2500 8845.1000 8845.2500 8890.1000 8890.2500 8891.1000 8891.2500 8892.1000 8892.2500 8898.1000 8898.2500 6282-01 6274-68 6274-02
J.T.Baker® HYDRA-POINT™, Volumetric Reagents for Water Determination by Karl Fischer titration
Karl Fischer-titrantit ja -reagenssit
No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free. They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples.
  • Pyridine-free
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
They are pyridine-free and specially formulated for existing application.