Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L

Toimittaja: Thermo Fisher Scientific

88632.LQ 88632.LS
88632.LQEA 247 EUR
88632.LQ 88632.LS
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
Standardiaineet ja vertailumateriaalit Ympäristöanalytiikan standardit ja vertailumateriaalit

CAS No.: 9005-84-9
Physical form: Liquid, for iodometric titrations
MDL No.: MFCD00082026

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR