Indium antimonidi ≥99.99% (metals basis), Electronic Grade

Toimittaja: Thermo Fisher Scientific
Varoitus

88460.09 88460.18 88460.04
88460.09EA 437 EUR
88460.09 88460.18 88460.04
Indium antimonidi ≥99.99% (metals basis), Electronic Grade
Indium antimonidi

Purity: 99.99% CAS No.: 1312-41-0 Molecular Formula: InSb Formula Weight: 236.57 Physical Form: Pieces Density: 5.75 at m.p. Melting Point: 535° Merck Reference: 14,4948 UN No.: 1549 MDL No.: MFCD00016146


In semiconductor electronics, as an infrared detector and filter

Kaava: InSb
Sulamispiste: 535 °C
Tiheys: 5,75 (m.p.
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016146
CAS-numero: 1312-41-0
EINECS: 215-192-3
UN: 1549
ADR: 6.1,III
Merck Index: 14,4948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR