Tioasetamidi ≥99.0% ACS for the precipitation of heavy metals, Fluka™

Toimittaja: Honeywell Chemicals
Vaara

Synonyms: Ethanethioamide

88450-250G 88450-50G 88450-1KG
88450-250GEA 449 EUR
88450-250G 88450-50G 88450-1KG
Tioasetamidi ≥99.0% ACS for the precipitation of heavy metals, Fluka™
Tioasetamidi

Thioacetamide ACS reagent for the precipitation of heavy metals.

Kaava: CH₃CSNH₂
MW: 75.13 g/mol
Sulamispiste: 108…114 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008070
CAS-numero: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity (IR) complying
Assay 99.0 - 101.0 %
Melting range 111 - 114 °C
Colour of the solution complying
Clearness of the solution complying
Residue on ignition (SO₄) Max. 0.05 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Application test complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR