Titanium nickel composite (70:30), jauhe -325 mesh

Toimittaja: Thermo Fisher Scientific
Vaara

88384.22 88384.36
88384.22EA 551 EUR
88384.22 88384.36
Titanium nickel composite (70:30), jauhe -325 mesh
Titanium nickel composite

Molecular Formula: Ti:Ni; 70:30 wt% Sensitivity: Air Sensitive UN No.: UN3089 MDL No.: MFCD02091734

Kaava: Ti:Ni
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02091734
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR