Sodium cyclopentadienide 2-3 M in THF

Toimittaja: Thermo Fisher Scientific
Vaara

88300.JQ 88300.JT
88300.JQEA 160 EUR
88300.JQ 88300.JT
Sodium cyclopentadienide 2-3 M in THF
Sodium cyclopentadienide

CAS No.: 4984-82-1 Molecular Formula: C5H5Na Formula Weight: 88.09 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.938 Sensitivity: Air and Moisture Sensitive Appearance: Pink to red Odour: Ether-like UN No.: UN3399 MDL No.: MFCD00012161


Applications: Reagent for the preparation of metallocenes

Kaava: C₅H₅Na
Tiheys: 0,938
Leimahduspiste: –17 °C (1 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012161
CAS-numero: 4984-82-1
EINECS: 225-636-8
UN: 3399
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR