Di-indiumtriselenidi ≥99.99% (metals basis)

Toimittaja: Thermo Fisher Scientific
Vaara

88280.06 88280.14
88280.06EA 382 EUR
88280.06 88280.14
Di-indiumtriselenidi ≥99.99% (metals basis)
Di-indiumtriselenidi

Purity: 99.99% CAS No.: 12056-07-4 Molecular Formula: In2Se3 Formula Weight: 466.52 Physical Form: 3-6mm Pieces Density: 5.67 Melting Point: ca 890° MDL No.: MFCD00016147

Kaava: In₂Se₃
Sulamispiste: ∼ 890 °C
Tiheys: 5,67
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016147
CAS-numero: 12056-07-4
EINECS: 235-016-9
UN: 3283
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR