Lantaani standardiliuos, 1000 mg/L La 5% typpihapossa (from La) Specpure® standard for AAS

Toimittaja: Thermo Fisher Scientific

88074.AEEA 125 EUR
88074.AE
Lantaani standardiliuos, 1000 mg/L La 5% typpihapossa (from La) Specpure® standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Lanthanum Standards

Molecular Formula: La in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066


Sertifikaatit: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR