Thallium(III) trifluoroacetate ≥95%

Toimittaja: Thermo Fisher Scientific
Vaara

87913.04 87913.09 87913.18
87913.04EA 101 EUR
87913.04 87913.09 87913.18
Thallium(III) trifluoroacetate ≥95%
Thallium(III) trifluoroacetate

Purity: 95% CAS No.: 23586-53-0 Molecular Formula: Tl(OOCCF3)3 Formula Weight: 543.42 Physical Form: Solid Sensitivity: Hygroscopic UN No.: 1707 MDL No.: MFCD00000414

Kaava: C₆F₉O₆Tl
Kiehumispiste: 160 °C (1,5 mmHg)
Sulamispiste: 213 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00000414
CAS-numero: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR