Kaliumkromaatti 5% aqueous solution Reag. Ph. Eur. 1069201

Toimittaja: VWR Chemicals
Vaara

87899.290EA 52.5 EUR
87899.290
Kaliumkromaatti 5% aqueous solution Reag. Ph. Eur. 1069201
Kaliumkromaatti

Tilaustiedot: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Kaava: K₂CrO₄
MW: 194.19 g/mol
Kiehumispiste: 1000
Sulamispiste: 985
MDL numero: MFCD00011368
CAS-numero: 7789-00-6
EINECS: 232-140-5
UN: 3082
ADR: 9 III
Merck Index: 12,07784

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR