Biureettireagenssiliuos Reag. Ph. Eur.

Toimittaja: VWR Chemicals
Varoitus

87787.290EA 158 EUR
87787.290
Biureettireagenssiliuos Reag. Ph. Eur.
Biureettireagenssiliuos

Tilaustiedot: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Storage Temperature: Huoneenlämpö
UN: 3266
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR