Organic reference standard, Erythrosin B, analytical standard, Supelco®
Toimittaja: Merck
87613-25MG
87613-25MGEA 149 EUR
87613-25MG
Organic reference standard, Erythrosin B, analytical standard, Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
Erythrosin B belongs to the class of xanthene dyes and has application as a food colorant.
It exhibits light absorption in the range of 500 - 550 nm which leads to the formation of reactive oxygen species using which it can inactivate the microorganisms, thereby acting like an anti-microbial agent.
Pakkaus: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906f3552ab0221c93c387466d5b6f0cd601fd7d9&vid=906f3552ab0221c93c387466d5b6f0cd601fd7d9&prid=web&clid=SAPDAM