Dimetyylisulfoksidi-D6 (99.8% D) for NMR spectroscopy

Toimittaja: VWR Chemicals


Filtered through a 0.2 µm filter, packaged under nitrogen.

87154.0006EA 70.9 EUR
87154.0006 87154.0007 87154.0025 87154.0010 87154.0100 87154.0011
Dimetyylisulfoksidi-D6 (99.8% D) for NMR spectroscopy
Dimetyylisulfoksidi-D6
Kaava: D₃CS(O)CD₃
Kiehumispiste: 189 °C (5 mmHg)
Tiheys: 1,19 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002090
CAS-numero: 2206-27-1
EINECS: 218-617-0

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.80 %
Water Max. 0.02 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR