Seleeni 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Toimittaja: VWR Chemicals
86709.260EA 45.7 EUR
86709.260 86709.180
Seleeni 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Selenium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%