Erbium 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Toimittaja: VWR Chemicals
86675.180EA 35.7 EUR
86675.180 86675.260
Erbium 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Erbium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%