Cesium 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Toimittaja: VWR Chemicals
86672.180EA 58 EUR
86672.180 86672.260
Cesium 1.000 mg/l laimeassa typpihapossa AVS TITRINORM standard for AAS
Standardiaineet ja vertailumateriaalit Cationic Standards Cesium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%