Typpihappo 10 mol/l (10 N), AVS TITRINORM

Toimittaja: VWR Chemicals
Vaara

Synonyms: Nitric acid solution

86172.290EA 77.4 EUR
86172.290
Typpihappo 10 mol/l (10 N), AVS TITRINORM
Typpihappo
Kaava: HNO₃
MW: 63.01 g/mol
Kiehumispiste: 87
Sulamispiste: -42
Tiheys: 1.51
MDL numero: MFCD00011349
CAS-numero: 7697-37-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Testitulosten erittely

Titer (20°C) 0.9980 – 1.0020
Method of analysis visual titration
Using sodium hydroxide volumetric solution
Indicator methyl red
Reference material indirect potassium hydrogenphthalate (KHP)
Traceability the reference material is a secondary reference material, traceable to standard material from NIST


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR