Fenolipunainen, AnalaR NORMAPUR® Reag. Ph. Eur., ACS indicator

Toimittaja: VWR Chemicals

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

86160.050EA 31.14 EUR
86160.050 86160.150 86160.180
Fenolipunainen, AnalaR NORMAPUR® Reag. Ph. Eur., ACS indicator
Fenolipunainen
Kaava: C₁₉H₁₄O₅S
MW: 354.38 g/mol
MDL numero: MFCD00003552
CAS-numero: 143-74-8
Merck Index: 13,07329

Order Now

Testitulosten erittely

pH Range (Yellow-Red) 6.8 - 8.2
Clarity of Solution Passes Test
Absorption Maximum λ1 (pH 1.2) 503 - 506
Absorption Maximum λ2 (pH 3.0) 430 - 435
Absorption Maximum λ3 (pH 6.5) 430 - 435
Absorption Maximum λ4 (pH 8.8) 557 - 560
Absorptivity (A1%/1 cm; λ1, on Dried Sample) 900 - 1100
Absorptivity (A1%/1 cm; λ2; on Dried Sample) 500 - 700
Absorptivity (A1%/1 cm; λ3, on Dried Sample) 500 - 700
Absorptivity (A1%/1 cm; λ4, on Dried Sample) 1000 - 1200
Insoluble in C₂H₅OH Passes Test
Copper (Cu) ≤0,005
Iron (Fe) ≤0,005
Lead (Pb) ≤0,005
Nickel (Ni) ≤0,005
Loss on Drying (110 °C) ≤5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR