Ksylenolioranssi, tetranatriumsuola, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator for metal titration

Toimittaja: VWR Chemicals

86150.010EA 57.3 EUR
86150.010 86150.050
Ksylenolioranssi, tetranatriumsuola, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. indicator for metal titration
Ksylenolioranssi, tetranatriumsuola
Kaava: C₃₁H₂₈N₂Na₄O₁₃S
Sulamispiste: ∼210 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00044293
CAS-numero: 3618-43-7
EINECS: 222-805-8

Order Now

Testitulosten erittely

Appearance of solution Passes test
Absorption maximum l (pH 14,0) 582 - 585 nm
Absortivity (A1%, 1cm, lmax, pH 14,0, on dried sample) 600 - 650
Loss on drying (110 ºC) Max. 7%
Suitability as indicator for metal titration Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR