Swabs for monitoring of surface, Contam
Toimittaja: LIOFILCHEM
86102 86100 86103 86101 86105 86104
86102.EA 155 EUR
86102. 86100. 86103. 86101. 86105. LIOF86104
Swabs for monitoring of surface, Contam
Valvontajärjestelmät Hygieniavalvonta
For microbial monitoring of pathogens on surfaces.
  • Easy to use
  • Clear colour changes
  • No additional reagents
  • Results in 18/24 hours
  • 10 CFU/ml detection limit