Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

860867-10G 860867-25G
860867-10GEA 110 EUR
860867-10G 860867-25G
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Brilliant cresyl blue

Brilliant cresyl blue ALD has been used for the staining of reticulocytes.


The structure of Brilliant Cresyl Blue ALD contains a 2-methyl modification on the phenoxazine core, while the structure of the historic Brilliant Cresyl Blue contains a 8-methyl modification on the phenoxazine core. The biological staining application is the same for both structures.

Kaava: C₃₄H₄₂Cl₄N₈O₂Zn
MDL numero: MFCD00148901
CAS-numero: 81029-05-2
EINECS: 279-675-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ba87fe9d53dcd67d47b0a617a0702325dd9d979&vid=68985ea7723e1e95168fb568a6de3ba76db42f89&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Dye content ≥50%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR