1,3-Dimethyl-2-imidazolidinone ≥99%, GPR RECTAPUR®

Toimittaja: VWR Chemicals
Vaara

85996.260EA 241 EUR
85996.260 85996.180
1,3-Dimethyl-2-imidazolidinone ≥99%, GPR RECTAPUR®
1,3-Dimethyl-2-imidazolidinone
Kaava: C₅H₁₀N₂O
MDL numero: MFCD00003188
CAS-numero: 80-73-9
UN: 2810
ADR: 6.1,III

Order Now

Testitulosten erittely

Assay Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR