Tioasetamidi 4% aqueous solution Reag. Ph. Eur. 1089602

Toimittaja: VWR Chemicals
Vaara

Synonyms: Ethanethioamide

85975.290EA 216 EUR
85975.290 85975.180 85975.260
Tioasetamidi 4% aqueous solution Reag. Ph. Eur. 1089602
Tioasetamidi

Tilaustiedot: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Kaava: CH₃CSNH₂
MW: 75.13 g/mol
Kiehumispiste: ~100 °C (1013 hPa)
Tiheys: 1 g/cm³ (20 °C)
MDL numero: MFCD00008070
CAS-numero: 62-55-5
EINECS: 200-541-4
Merck Index: 13,09391
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2000e62db991c85340c257ac6d64ca42c3551a&vid=18e76b2cdee9ee22a2618ec371858a498f218457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR