Kloorivetyliuos 2 mol/l (2 N) Reag. Ph. Eur. 1043503

Toimittaja: VWR Chemicals
Varoitus

85848.290EA 112 EUR
85848.290
Kloorivetyliuos 2 mol/l (2 N) Reag. Ph. Eur. 1043503
Kloorivetyliuos

Tilaustiedot: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Kaava: HCl
Tiheys: 1,031 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR