VWR®, N,N-Dimethylformamide, secondary reference standard for GC, PESTINORM®
Toimittaja: VWR Chemicals
85662.180EA 248 EUR
85662.180
VWR®, N,N-Dimethylformamide, secondary reference standard for GC, PESTINORM®
Standardiaineet ja vertailumateriaalit Kaasukromatografiastandardit
A secondary GC solvent reference standard for various chromatographic and analytical applications.
  • Purity and the specific impurity profile is determined by GC-MS and reported in the CoA
  • Traceability data to specific current lot of primary standard
  • Completely synthetic and typically over 99,9% pure by GC-FID