ARISTAR®, Erbium Standard Solutions for ICP-MS
Toimittaja: VWR Chemicals
85563.180EA 86.4 EUR
85563.180 457257G
ARISTAR®, Erbium Standard Solutions for ICP-MS
Standardiaineet ja vertailumateriaalit Cationic Standards Erbium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Sertifikaatit: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Limited Time Sale
Select items below are currently on sale