Sitruunahappo monohydraatti 99.5-100.5%, jauhe Ph. Eur.

Toimittaja: VWR Chemicals
Varoitus

85514.290EA 96.2 EUR
85514.290 85514.360 85514.460
Sitruunahappo monohydraatti 99.5-100.5%, jauhe Ph. Eur.
Sitruunahappomonohydraatii

Varoitus: Not suitable for parenteral use

Kaava: C₆H₈O₇.H₂O
Sulamispiste: 135…152 °C
Tiheys: 1,552 g/cm³ (20 °C)
Leimahduspiste: 173,9 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149972
CAS-numero: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (calculated on anhydrous) 99.5 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification B Passes test
Appearance of solution Passes test
Readily carbonisable substances Passes test
Oxalic acid (calculated on anhydrous) Max. 360 ppm
SO4 (Sulphate) Max. 150 ppm
Heavy metals (as Pb) Max. 10 ppm
Water 7.5 - 9.0 %
Sulphated ash Max. 0.1 %
Residual solvents Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR