VWR®, Coulometric reagents for water determination by the Karl Fischer method
Toimittaja: VWR Chemicals
85470.290EA 223 EUR
85470.290 85471.130 85472.260 85473.260 85474.260 85475.260 85476.260 85477.260 85478.260 85479.130
VWR®, Coulometric reagents for water determination by the Karl Fischer method
Karl Fischer-titrantit ja -reagenssit
Both anolyte and catholyte solutions are with improved formulation to increase the speed and accuracy of titration when determining water content at the microgram level.
  • Reach endpoint quickly and consistently
  • Long shelf life
  • Stable endpoint