Test strips, peroxide, Dosatest
Toimittaja: VWR Chemicals
85433.601EA 31.3 EUR
85433.601 85434.601 85435.601
Test strips, peroxide, Dosatest
Ympäristöanalytiikan pikatestit
A ready to use kit that fits into your lab coat pocket. Intended for water-based samples
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Pakkaus: Container with 100 test strips 6×95 mm.