Bariumkloridi in aqueous solution USP Test Solution (TS)

Toimittaja: VWR Chemicals

85332.180EA 45.1 EUR
85332.180 85332.260
Bariumkloridi in aqueous solution USP Test Solution (TS)
Bariumkloridi

Ready to use test/reagent solutions (TS and RS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Kaava: BaCl₂
MW: 208.24 g/mol
Kiehumispiste: 1560
Sulamispiste: 963
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003445
CAS-numero: 10361-37-2
EINECS: 0
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR