Dimetyylidikloorisilaani 5% in heptane Silanization solution I, Selectophore™ ionophore, Supelco®

Toimittaja: Merck
Vaara


M-Clarity™ quality level = MQ100

85126-1L 85126-250ML
85126-1LEA 227 EUR
85126-1L 85126-250ML
Dimetyylidikloorisilaani 5% in heptane Silanization solution I, Selectophore™ ionophore, Supelco®
Dimetyylidikloorisilaani

Ready-to-use solution for silanizing micro-electrodes.

Kaava: C₂H₆Cl₂Si
MDL numero: MFCD00000491
CAS-numero: 75-78-5
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29a64e31328c65f3e1562b829e50639e996c41d&vid=0a4419ea13f3484b5717c588fe35e543f9fcbec9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR