VWR®, Refractive index standards
Toimittaja: VWR Chemicals
85133.015EA 91 EUR
85133.015 85134.015 85135.015 85136.015 85137.015 85138.015 85114.015 85115.015 85116.015 85117.015 85118.015 85119.015 85120.015 85121.015 85122.015 85123.015 85124.015 85125.015 85126.015 85127.015 85128.015 85129.015 85130.015 85131.015 85132.015
VWR®, Refractive index standards
Standardiaineet ja vertailumateriaalit Taitekerroinstandardit
Refractive index of each standard is verified using a calibrated and temperature controlled refractometer.
  • Standards are prepared gravimetrically on weight/weight basis
  • Solute (sucrose) and solvent (water) are weighed on a calibrated balance using OIML traceable weights
Sertifikaatit: Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.