VWR®, COD calibration standards
Toimittaja: VWR Chemicals
85050.260EA 154 EUR
85050.260 85051.260 85052.260 85053.260 85054.260 85055.260 85056.260 85057.260 85058.260 85059.260 85060.260 85061.260 85062.260 85063.260 85064.260 85065.260 85066.260
VWR®, COD calibration standards
Standardiaineet ja vertailumateriaalit Ympäristöanalytiikan standardit ja vertailumateriaalit
These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.
Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.