Rikkihappo 93% ≥93%, NORMATOM®, Ultrapure for trace metal analysis

Toimittaja: VWR Chemicals
Vaara

85028.270EA 544.2 EUR
85028.270 85028.290 85028.230
Rikkihappo 93% ≥93%, NORMATOM®, Ultrapure for trace metal analysis
Rikkihappo
Kaava: H₂SO₄
MW: 98.08 g/mol
Kiehumispiste: 330
Sulamispiste: 10.38
Tiheys: 1.82 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064589
CAS-numero: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Testitulosten erittely

Assay w/w 93-98%
Ag (Silver) 50 ppt
Al (Aluminium) 50 ppt
As (Arsenic) 500 ppt
Ba (Baryum) 10 ppt
Be (Beryllium) 10 ppt
Bi (Bismuth) 10 ppt
Ca (Calcium) 50 ppt
Cd (Cadmium) 10 ppt
Ce (Cerium) 10 ppt
Co (Cobalt) 10 ppt
Cr (Chromium) 10 ppt
Cs (Cesium) 10 ppt
Cu (Copper) 10 ppt
Dy (Dysprosium) 10 ppt
Er (Erbium) 10 ppt
Eu (Europium) 10 ppt
Fe (Iron) 50 ppt
Ga (Gallium) 10 ppt
Gd (Gadolinium) 10 ppt
Ge (Germanium) 100 ppt
Hf (Hafnium) 10 ppt
Hg (Mercury) 100 ppt
Ho (Holmium) 10 ppt
In (Indium) 10 ppt
K (Potassium) 50 ppt
La (Lanthanum) 10 ppt
Li (Lithium) 10 ppt
Lu (Lutetium) 10 ppt
Mg (Magnesium) 50 ppt
Mn (Manganese) 10 ppt
Mo (Molybdenum) 10 ppt
Na (Sodium) 50 ppt
Nb (Niobium) 10 ppt
Nd (Neodymium) 10 ppt
Ni (Nickel) 50 ppt
Pb (Lead) 10 ppt
Pd (Palladium) Information only
Pr (Praseodymium) 10 ppt
Pt (Platinum) Information only
Rb (Rubidium) 10 ppt
Rh (Rhodium) 50 ppt
Sb (Antimony) 50 ppt
Sc (Scandium) 10 ppt
Se (Selenium) 500 ppt
Sm (Samarium) 10 ppt
Sn (Tin) 50 ppt
Sr (Strontium) 10 ppt
Ta (Tantalum) Information only
Tb (Terbium) 10 ppt
Te (Tellurium) 50 ppt
Th (Thorium) 10 ppt
Ti (Titanum) 50 ppt
Tl (Thallium) 10 ppt
Tm (Thulium) 10 ppt
U (Uranium) 10 ppt
V (Vanadium) 10 ppt
W (Tungsten) 10 ppt
Y (Yttrium) 10 ppt
Yb (Ytterbium) 10 ppt
Zn (Zinc) 50 ppt
Zr (Zirconium) 10 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR