Trifluorietikkahappo ≥99.9%, HiPerSolv CHROMANORM® for LC-MS

Toimittaja: VWR Chemicals
Vaara

Synonyms: TFA

Packaged under nitrogen.

84868.180EA 601 EUR
84868.180 84868.260 84868.290
Trifluorietikkahappo ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Trifluorietikkahappo
Kaava: C₂HF₃O₂
MW: 114.02 g/mol
Kiehumispiste: 71.8 °C (1013 hPa)
Sulamispiste: –15 °C
Tiheys: 1.490 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004169
CAS-numero: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.9 %
Evaporation residue Max. 0.0001 %
Water Max. 0.005 %
Ca (Calcium) (0.1 %) Max. 0.2 ppm
K (Potassium) (0.1 %) Max. 0.2 ppm
Mg (Magnesium) (0.1 %) Max. 0.2 ppm
Na (Sodium) (0.1 %) Max. 0.5 ppm
Transmittance (205 nm) (0.1 %) Min. 10 %
Transmittance (220 nm) (0.1 %) Min. 50 %
Transmittance (230 nm) (0.1 %) Min. 80 %
Transmittance (250 nm) (0.1 %) Min. 95 %
Transmittance (255 nm) (0.1 %) Min. 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR