VWR®, Colour standard solutions ASTM
Toimittaja: VWR Chemicals
84796.180EA 85.2 EUR
84796.180 84796.260 84797.180 84797.260 84798.260 84834.180 84834.260
VWR®, Colour standard solutions ASTM
Standardiaineet ja vertailumateriaalit Colour Reference and Spectroscopy Standards
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis.
  • Use either as calibration and/or quality control standards
  • Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
  • Traceable product certified by an independent laboratory
  • Supplied in high quality, tamper-evident bottles