o-Cresolphthalein 20 g/l in ethanol 70%, VOLUSOL®

Toimittaja: VWR Chemicals
Vaara

84599.180EA 93.7 EUR
84599.180 84599.260
o-Cresolphthalein 20 g/l in ethanol 70%, VOLUSOL®
o-Cresolphthalein

An alternative to phenolphthalein in solution.

Kaava: C₂₂H₁₈O₄
Tiheys: 0,886 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005912
CAS-numero: 596-27-0
EINECS: 209-881-8
UN: 1170
ADR: 3,III

Order Now

Testitulosten erittely

Transition range pH 8.2 - 9.8 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR